Temperature chart recorder
Leverandør: Thermo Scientific
6383-6
HERA6383-6EA 34775 NOK
HERA6383-6
Temperature chart recorder
Punktskriver Skrivere
These recorders are ideal when ultra-low temperature freezers need to meet validation and regulatory requirements.
  • Record seven-day history on 150 mm charts with pressure sensitive chart paper
  • No inking is required