Fiberlite™ Rotor Adapters
Leverandør: Thermo Scientific
75100378
HERA75100378EA 3000 NOK
HERA75100378
Fiberlite™ Rotor Adapters
Sentrifuger Rotorer
Incorporate Thermo Scientific™ Fiberlite™ Rotor Adapters with Fiberlite F13-14x50c and F15-8x50c rotors.