High-Performance Chromatography Refrigerators, TSX
Leverandør: Thermo Scientific
TSX3005CV TSX5005CV TSX2305CV
HERATSX3005CVEA 0 NOK
HERATSX3005CV HERATSX5005CV HERATSX2305CV
High-Performance Chromatography Refrigerators, TSX
Kjøleskap
The TSX high-performance chromatography refrigerators are designed for a variety of applications requiring close temperature control, full access to chromatography instrumentation, and easy set-up of instrumentation and apparatus within the chamber. The TSX chromatography series features industry-leading safety features to ensure maximum protection.
  • Forced-air circulation for temperature uniformity at all shelf levels and Heat-free defrost for maximum temperature uniformity
  • Quiet operation at just 52 dBA
  • Two 50 mm side wall access ports with insulated