High-Performance Undercounter Lab Refrigerators, TSX
Leverandør: Thermo Scientific
TSX505GA TSX505SA
HERATSX505GAEA 66850 NOK
HERATSX505GA HERATSX505SA
High-Performance Undercounter Lab Refrigerators, TSX
Frysere Frysere, generelt bruk
Thermo Scientific™ TSX series high-performance undercounter lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, pharmaceuticals, vaccines, chemotherapy, and other medical and pharmacy-grade storage requiring 2 to 8 °C. The series V-drive is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
  • Designed to safely store media, reagents, pharmaceuticals, vaccines,