Swabs in tubes
Leverandør: HERENZ HEINZ
1020001 1020002 1020003 1020025
HERE1020001EA 3475 NOK
HERE1020001 HERE1020002 HERE1020003 HERE1020025
Swabs in tubes
Svabere Svabere i rør
Sterile dry swabs in tubes, without transport medium.
  • Handle is made of PS, aluminium or wood
  • Head is made of viscose/rayon or cotton
  • Unbreakable PP tube (approx. 150×12 mm)
With label on the tube and stopper.
Forpakning: Available loose or individually packed in a peel pack.