Alcohol swab
Leverandør: HERENZ HEINZ
1131897
HERE1131897EA 2400 NOK
HERE1131897
Alcohol swab
Svabere Svabere for rengjøring
70% isopropyl alcohol non-sterile swabs for cleaning applications.
Unfolded size of the swab is approximately 60×30 mm.
Forpakning: Individual – 100 pieces per box.