Avantor® Hichrom HI-1 MS, GC Columns
Leverandør: Avantor
HI16-25-025-15G5EA 7325 NOK
HI16-25-025-15G5 HI16-25-025-30G5 HI16-25-025-60G5 HI16-53-100-30G5 HI16-18-018-10 HI16-18-018-50 HI16-18-018-60 HI16-18-020-20 HI16-18-020-40 HI16-18-040-10 HI16-18-040-20 HI16-18-040-60 HI16-20-010-10 HI16-20-010-60 HI16-20-020-10 HI16-20-020-40 HI16-20-033-12 HI16-20-033-50 HI16-20-040-20 HI16-20-040-40 HI16-20-040-50 HI16-20-100-20 HI16-20-100-40 HI16-25-010-30 HI16-25-010-60 HI16-25-025-105 HI16-25-025-15 HI16-25-025-50 HI16-25-050-10 HI16-25-050-105 HI16-25-050-15 HI16-25-050-60 HI16-25-100-105 HI16-32-010-105 HI16-32-025-105 HI16-32-025-15 HI16-32-025-30 HI16-32-025-50 HI16-32-050-105 HI16-32-050-60 HI16-32-100-105 HI16-32-100-50 HI16-32-150-10 HI16-32-150-105 HI16-32-150-15 HI16-32-150-50 HI16-53-010-10 HI16-53-010-15 HI16-53-025-15 HI16-53-050-10 HI16-53-100-10 HI16-53-100-15 HI16-53-100-30 HI16-53-100-75 HI16-53-150-50 HI16-05-005-2.5 HI16-10-010-10 HI16-10-010-5 HI16-10-020-10 HI16-10-020-15 HI16-10-040-15 HI16-10-040-5 HI16-15-015-20 HI16-15-015-30 HIGCHI1002502515
Avantor® Hichrom HI-1 MS, GC Columns
Kromatografikolonner
Really versatile general purpose apolar phase for a wide range of MS applications, including (for example): PCBs, simulated distillation, gases, essential oils, hydrocarbons analysis, pesticides, semivolatiles, oxygenates. HI-1 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
  • Phase composition: Crossbonded 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - MS grade
  • Long lifetime
  • Equivalent to USP phases G1, G2, G38
Max temperature limits may change depending on the film thickness.