Avantor® Hichrom HI-PS264, GC Columns
Leverandør: Avantor
HI59-15-004-10EA 7975 NOK
HI59-15-004-10 HI59-18-018-10 HI59-18-018-20 HI59-18-018-40 HI59-18-018-50 HI59-18-018-60 HI59-18-020-10 HI59-18-020-20 HI59-18-020-40 HI59-18-040-10 HI59-18-040-20 HI59-18-040-40 HI59-18-040-50 HI59-18-040-60 HI59-20-010-10 HI59-20-010-20 HI59-20-010-40 HI59-20-010-50 HI59-20-010-60 HI59-20-020-10 HI59-20-020-20 HI59-20-020-40 HI59-20-020-50 HI59-20-020-60 HI59-20-040-10 HI59-20-040-20 HI59-20-040-40 HI59-20-040-50 HI59-20-040-60 HI59-20-100-10 HI59-20-100-20 HI59-20-100-40 HI59-20-100-50 HI59-20-100-60 HI59-25-010-15 HI59-25-010-30 HI59-25-010-50 HI59-25-010-60 HI59-25-025-10 HI59-25-025-105 HI59-25-025-15 HI59-25-025-30 HI59-25-025-50 HI59-25-025-60 HI59-25-050-10 HI59-25-050-105 HI59-25-050-15 HI59-25-050-30 HI59-25-050-50 HI59-25-050-60 HI59-25-100-10 HI59-25-100-105 HI59-25-100-15 HI59-25-100-30 HI59-25-100-50 HI59-25-100-60 HI59-25-200-10 HI59-25-200-105 HI59-25-200-15 HI59-25-200-30 HI59-25-200-50 HI59-25-200-60 HI59-25-300-10 HI59-25-300-15 HI59-25-300-30 HI59-25-300-50 HI59-25-300-60 HI59-32-010-10 HI59-32-010-105 HI59-32-010-15 HI59-32-010-30 HI59-32-010-50 HI59-32-010-60 HI59-32-025-10 HI59-32-025-105 HI59-32-025-15 HI59-32-025-30 HI59-32-025-50 HI59-32-025-60 HI59-32-050-10 HI59-32-050-105 HI59-32-050-15 HI59-32-050-30 HI59-32-050-50 HI59-32-050-60 HI59-32-100-10 HI59-32-100-105 HI59-32-100-15 HI59-32-100-30 HI59-32-100-50 HI59-32-100-60 HI59-32-150-10 HI59-32-150-105 HI59-32-150-15 HI59-32-150-30 HI59-32-150-50 HI59-32-150-60 HI59-32-300-10 HI59-32-300-105 HI59-32-300-15 HI59-32-300-30 HI59-32-300-50 HI59-32-300-60 HI59-32-400-30 HI59-32-500-10 HI59-32-500-105 HI59-32-500-15 HI59-32-500-30 HI59-32-500-50 HI59-32-500-60 HI59-53-010-10 HI59-53-010-15 HI59-53-010-30 HI59-53-025-15 HI59-53-025-30 HI59-53-025-50 HI59-53-025-60 HI59-53-050-10 HI59-53-050-15 HI59-53-050-30 HI59-53-050-50 HI59-53-050-60 HI59-53-100-10 HI59-53-100-15 HI59-53-100-30 HI59-53-100-50 HI59-53-100-60 HI59-53-100-75 HI59-53-150-10 HI59-53-150-15 HI59-53-150-30 HI59-53-150-50 HI59-53-150-60 HI59-53-150-75 HI59-53-265-10 HI59-53-300-10 HI59-53-300-15 HI59-53-300-30 HI59-53-300-50 HI59-53-300-60 HI59-53-300-75 HI59-53-500-10 HI59-53-500-15 HI59-53-500-30 HI59-53-500-50 HI59-53-500-60 HI59-53-500-75 HI59-53-700-15 HI59-53-700-30 HI59-05-005-2 HI59-05-005-2.5 HI59-05-005-5 HI59-10-010-10 HI59-10-010-15 HI59-10-010-5 HI59-10-020-10 HI59-10-020-15 HI59-10-020-5 HI59-10-040-10 HI59-10-040-15 HI59-10-040-5 HI59-15-015-10 HI59-15-015-15 HI59-15-015-20 HI59-15-015-30 HI59-15-015-5 HI59-15-004-3G2 HI59-25-025-15G5 HI59-25-025-30G5 HI59-25-025-60G5 HI59-53-100-30G5
Avantor® Hichrom HI-PS264, GC Columns
Kromatografikolonner
Apolar phase, suitable for high flm thickness columns, to analyze solvents, alcohols, volatiles, etc.
Max temperature limits may change depending on the film thickness.