Avantor® Hichrom MicroTight®, Fittings
Leverandør: Avantor
HICHHI-799EA 1725 NOK
HICHHI-799
Avantor® Hichrom MicroTight®, Fittings
Fittings og kontakter
MicroTight® one-piece finger tight PEEK™ fittings are used to connect capillary tubing to MicroTight® unions, adapters or filters.
  • Pressure rated to 276 bar
  • Temperature stability: 125 °C