MIU medium base dehydrated culture media
Leverandør: HIMEDIA
M1076-500G
HIMEM1076-500GEA 2050 NOK
HIMEM1076-500G
MIU medium base dehydrated culture media
Mikrobiologiske medier Media, dehydrert for mikrobiologi
For the detection of motility and urease and indole production.
  • Microbiological culture media
  • 500 g poly bottles