Sedimenteringskjegler, Imhoff
Leverandør: HIRSCHMANN
7720193
HIRS7720193EA 2625 NOK
HIRS7720193
Sedimenteringskjegler, Imhoff
Trakter Immhoff-trakter
Sedimentation cone, Imhoff, made of SAN, clear and unbreakable with elevated scale, screw cap for rapid emptying.
  • As transparent as glass
  • Easy draining via screw plug
Graduation: 0 to 2 ml:0,1/ 2 to 10 ml:0,5/ 10 to 40 ml:1/ 40 to 100 ml:2/ 100 to 1000 ml:10