Byrettestoppekraner, rett ventil
Leverandør: HIRSCHMANN
8901060
HIRS8901060EA 740 NOK
HIRS8901060
Byrettestoppekraner, rett ventil
Haner
DURAN®, borosilicate glass, clear, with PTFE spindle.
  • Excellent chemical resistance
  • Excellent temperature resistance
Straight for burettes.