HPLC columns, Develosil® ODS-UG
Leverandør: HICHROM
UG11546250W
554-7681EA 0 NOK
554-7681 554-7680 HISMUG11546250W
HPLC columns, Develosil® ODS-UG
Kromatografikolonner
Develosil® ODS-UG is the most stable column under alkaline conditions (pH 1 to 10).
  • Spherical porous silica