VWR® Hitachi, Spare parts for HPLC system, Chromaster
Leverandør: VWR Collection
HITA893-0809EA 10250 NOK
HITA893-0809
VWR® Hitachi, Spare parts for HPLC system, Chromaster
Kromatografiutstyr Kromatografireservedeler
This spare part is suitable for Chromaster 5210 and 5260 autosamplers. For Chromaster 5210 AS from Serial No. 1309-001 and 1309T-001 onward. For Chromaster 5260 AS from Serial No. 1205-001 and 1205T-001 onward.