ESD cleanroom shoe covers, anti-slip
Leverandør: HUBEI ORIENT INTL TRADING
HOIT110228EA 4900 NOK
HOIT110228
ESD cleanroom shoe covers, anti-slip
Engangsplagg
The anti-static ESD shoe covers are manufactured with fabric strips.
  • Skid-free soles for extra traction and improved safety
  • Made of PP
  • Non sterile
Weight: 37 g/m²
Forpakning: 100 pieces/bag, 3 bags/case.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Kjønn
Antrekksapplikasjoner Cleanroom