Laboratory coats, PP
Leverandør: HUBEI ORIENT INTL TRADING
HOIT115453EA 1425 NOK
HOIT115453
Laboratory coats, PP
Engangsplagg
Soft and comfortable, this non-woven coat is perfect for dusty areas.
  • Low lint properties, and provides excellent splash protection
  • High-tech fabric that offers cloth-like comfort that is easy to wear
  • With elastic wrist, without pockets and shirt neck
  • Zip for closure
Greater range of movement, improved mobility with more room in the back and reinforcement in high stress areas for fewer blowouts.
Forpakning: 5pcs/polybag; 10bags/carton.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Kjønn Unisex
Antrekksapplikasjoner Laboratory