EDTA dinatrium salt dihydrat 99.0-101.0%, meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™

Leverandør: Honeywell Chemicals
Warning

27285H-1KG 27285H-500G
HONC27285H-1KGEA 4075 NOK
HONC27285H-1KG HONC27285H-500G
EDTA dinatrium salt dihydrat 99.0-101.0%, meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Identity (IR) complying
Assay (calculated to the dried substance) 99.0 - 101.0 %
Loss on drying (150°C) 8.7 - 11.4 %
pH (1 %,25°C) 4.3 - 4.7
pH (5 %, 25 °C) 4.0 - 6.0
pH (5 %, 20°C) 4.0 - 5.0
Arsenic (As) Max. 0.0001 %
Calcium (Ca) complying
Iron (Fe) Max. 10 ppm
Lead (Pb) Max. 10 ppm
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 0.01 %
Cyanide (CN) Max. 0.001 %
Sulfate (SO₄) Max. 0.05 %
Nitrilotriacetic acid (HPLC) Max. 0.1 %
Residual solvents complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR