Indigo carmine solution for PANPEHA indicator paper, Riedel-de Haën™

Leverandør: Honeywell Chemicals

Synonymer: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue

34661-2.5L
HONC34661-2.5LEA 7675 NOK
HONC34661-2.5L
Indigo carmine solution for PANPEHA indicator paper, Riedel-de Haën™
Indigo carmine
Formel: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
MDL nummer: MFCD00005723
CAS-nummer: 860-22-0
Merck Indeks: 13,04969

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR