Aluminium ICP Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1004-100ML E1100-100ML E1131-100ML E1201-100ML
HONCE1004-100MLEA 920 NOK
HONCE1004-100ML HONCE1100-100ML HONCE1131-100ML HONCE1201-100ML
Aluminium ICP Standards, (CRMs), Fluka™
Standarder Kationstandarder Aluminium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).