Chromium ICP standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1015-100ML E1104-100ML E1142-100ML
HONCE1015-100MLEA 920 NOK
HONCE1015-100ML HONCE1104-100ML HONCE1142-100ML
Chromium ICP standards, (CRMs), Fluka™
Standarder Kationstandarder Chromium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).