Hafnium ICP Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1025-100ML E1222-100ML
HONCE1025-100MLEA 2700 NOK
HONCE1025-100ML HONCE1222-100ML
Hafnium ICP Standards, (CRMs), Fluka™
Standarder Kationstandarder Hafnium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).