Iridium ICP Standard, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1029-100ML
HONCE1029-100MLEA 3375 NOK
HONCE1029-100ML
Iridium ICP Standard, (CRMs), Fluka™
Standarder Kationstandarder Iridium Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).