Lanthanum ICP Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1031-100ML E1158-100ML
HONCE1031-100MLEA 920 NOK
HONCE1031-100ML HONCE1158-100ML
Lanthanum ICP Standards, (CRMs), Fluka™
Standarder Kationstandarder Lanthanum Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).