Europium ICP & ICP-MS Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1317-100ML E1385-100ML
HONCE1317-100MLEA 1325 NOK
HONCE1317-100ML HONCE1385-100ML
Europium ICP & ICP-MS Standards, (CRMs), Fluka™
Standarder Kationstandarder Europium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).