Indium ICP & ICP-MS standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1325-100ML E1393-100ML
HONCE1325-100MLEA 1325 NOK
HONCE1325-100ML HONCE1393-100ML
Indium ICP & ICP-MS standards, (CRMs), Fluka™
Standarder Kationstandarder Indium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).