Iridium ICP and ICP-MS Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1326-100ML
HONCE1326-100MLEA 2025 NOK
HONCE1326-100ML
Iridium ICP and ICP-MS Standards, (CRMs), Fluka™
Standarder Kationstandarder Iridium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).