Bismuth ICP and ICP-MS standard, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1368-100ML
HONCE1368-100MLEA 1125 NOK
HONCE1368-100ML
Bismuth ICP and ICP-MS standard, (CRMs), Fluka™
Standarder Kationstandarder Bismuth Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).