Beryllium ICP & ICP-MS Standard, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1375-100ML
HONCE1375-100MLEA 1775 NOK
HONCE1375-100ML
Beryllium ICP & ICP-MS Standard, (CRMs), Fluka™
Standarder Kationstandarder Beryllium Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).