Gallium ICP & ICP-MS Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1319-100ML E1387-100ML
HONCE1319-100MLEA 1150 NOK
HONCE1319-100ML HONCE1387-100ML
Gallium ICP & ICP-MS Standards, (CRMs), Fluka™
Standarder Kationstandarder Gallium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).