Sodium ICP & ICP-MS Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E1334-100ML E1402-100ML
HONCE1334-100MLEA 1150 NOK
HONCE1334-100ML HONCE1402-100ML
Sodium ICP & ICP-MS Standards, (CRMs), Fluka™
Standarder Kationstandarder Sodium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).