Combined Seven Anion Standard I for IC
Leverandør: Honeywell Chemicals
E2029-100ML
HONCE2029-100MLEA 2875 NOK
HONCE2029-100ML
Combined Seven Anion Standard I for IC
Standarder Anionstandarder Multi-Element Anioinic Standards
Certified Reference Material (CRM).
150 mg/L of PO43-, SO42-; 100 mg/L of NO2-, Br-, NO3-; 30 mg/L Cl-; 20 mg/L F- in H₂O