Combined Six Cation Standard I for IC
Leverandør: Honeywell Chemicals
E2032-100ML
HONCE2032-100MLEA 2875 NOK
HONCE2032-100ML
Combined Six Cation Standard I for IC
Standarder Anionstandarder Multi-Element Anioinic Standards
Certified Reference Material (CRM).
1000 mg/L Ca2+, 400 mg/L NH4+, 200 mg/L each of Na+, K+, Mg2+, 50 mg/L Li+ Material in H₂O