Combined Six Cation Standard II for IC
Leverandør: Honeywell Chemicals
E2033-100ML
HONCE2033-100MLEA 2875 NOK
HONCE2033-100ML
Combined Six Cation Standard II for IC
Standarder Anionstandarder Multi-Element Anioinic Standards
Certified Reference Material (CRM).
500 mg/L each of K+, Ca2+, 250 mg/L each of NH4+, Mg2+, 200 mg/L Na+, 50 mg/L Li+ in H₂O