Mixed calibration standard 4 for ICP
Leverandør: Honeywell Chemicals
E2097-100ML
HONCE2097-100MLEA 4800 NOK
HONCE2097-100ML
Mixed calibration standard 4 for ICP
Standarder Kationstandarder Multi-Element Standards
Certified Reference Material (CRM).
1000 mg/L Ca, 400 mg/L K, 200 mg/L each of Al, Na, 20 mg/L each of Cr, Ni in HNO₃ 5%