Concentrated Eluent for Ion Chromatography, Sodium Carbonate Solution
Leverandør: Honeywell Chemicals
E2206-500ML
HONCE2206-500MLEA 2775 NOK
HONCE2206-500ML
Concentrated Eluent for Ion Chromatography, Sodium Carbonate Solution
Standarder Anionstandarder Multi-Element Anioinic Standards
Certified Reference Material (CRM).
0.5 M Sodium Carbonate (Na₂CO₃) in water