Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 2
Leverandør: Honeywell Chemicals
E2208-500ML
HONCE2208-500MLEA 2775 NOK
HONCE2208-500ML
Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 2
Standarder Anionstandarder Multi-Element Anioinic Standards
Certified Reference Material (CRM).
0.18 M Na₂CO₃ / 0.17 M NaHCO₃ in water