Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 3
Leverandør: Honeywell Chemicals
E2209-500ML
HONCE2209-500MLEA 2775 NOK
HONCE2209-500ML
Concentrated Eluent for Ion Chromatography, Sodium Carbonate/Sodium Bicarbonate Solution 3
Standarder Anionstandarder Multi-Element Anioinic Standards
Certified Reference Material (CRM).
0.35 M Na₂CO₃ / 0.10 M NaHCO₃ in water