Chromium(VI) standard for ion chromatography, (CRM), Fluka™
Leverandør: Honeywell Chemicals
E3012-100ML
HONCE3012-100MLEA 815 NOK
HONCE3012-100ML
Chromium(VI) standard for ion chromatography, (CRM), Fluka™
Standarder Kationstandarder Chromium Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).