Methansulfonate standard for ion chromatography, (CRM), Fluka™
Leverandør: Honeywell Chemicals
E3027-100ML
HONCE3027-100MLEA 765 NOK
HONCE3027-100ML
Methansulfonate standard for ion chromatography, (CRM), Fluka™
Standarder Anionstandarder Methansulphonate Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).