Nitrilotriacetate standard for ion chromatography, (CRM), Fluka™
Leverandør: Honeywell Chemicals
E3033-100ML
HONCE3033-100MLEA 1025 NOK
HONCE3033-100ML
Nitrilotriacetate standard for ion chromatography, (CRM), Fluka™
Standarder Anionstandarder Nitrilotriacetate Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).