Perchlorate Standard for Ion Chromatography, (CRM), Fluka™
Leverandør: Honeywell Chemicals
E3038-100ML
HONCE3038-100MLEA 620 NOK
HONCE3038-100ML
Perchlorate Standard for Ion Chromatography, (CRM), Fluka™
Standarder Anionstandarder Perchlorate Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).