Sodium Standard for Ion Chromatography, (CRM), Fluka™
Leverandør: Honeywell Chemicals
E3044-100ML
HONCE3044-100MLEA 645 NOK
HONCE3044-100ML
Sodium Standard for Ion Chromatography, (CRM), Fluka™
Standarder Kationstandarder Sodium Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).