Samarium AAS Standards, (CRMs), Fluka™
Leverandør: Honeywell Chemicals
E4069-100ML E4069-500ML
HONCE4069-100MLEA 640 NOK
HONCE4069-100ML HONCE4069-500ML
Samarium AAS Standards, (CRMs), Fluka™
Standarder Kationstandarder Samarium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Sertifiseringer: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).