Medical Masks, Type IIR
Leverandør: Medicom
M36101-40
HOPEM36101-40EA 1675 NOK
HOPEM36101-40
Medical Masks, Type IIR
Masker Munnbind, kirurgi
Medical Device: Class I - Medical mask type: Type IIR - Elastic.
  • Protects the environment from droplets emitted by the wearer while providing protection against splashes of biological fluids
  • Effective against bacteria, particles and viruses (BFE, PFE, & VFE tested)
  • Kolmi masks feature the exclusive Origami folding and Smile welding for greater comfort and a perfect fit to the wearer's face
  • Hypoallergenic