Pocket salt meter, ISE method, LAQUAtwin Salt-22
Leverandør: HORIBA
3200689158
HORI3200689158EA 8125 NOK
HORI3200689158
Pocket salt meter, ISE method, LAQUAtwin Salt-22
Elektrokjemiske målere Salinitetsmåler
This meter directly measures sodium ion concentration in 0,3 ml sample (or 0,05 ml with sampling sheet B) and converts it into salinity or NaCl salt concentration. The replaceable flat sensor utilises a sodium ion selective electrode (ISE) that requires only a few drops to enable quick and accurate analysis.
  • Automatic calibration and standard recognition
  • Automatic hold / auto stable
  • Durable waterproof ABS housing, LCD display with backlight
  • Auto power off after 30 mins, low battery indicator
  • 400 hours battery life (continuous use without backlight)
  • No need