Pocket salt meter, EC-based, LAQUAtwin Salt-11
Leverandør: HORIBA
3999960128
HORI3999960128EA 3125 NOK
HORI3999960128
Pocket salt meter, EC-based, LAQUAtwin Salt-11
Elektrokjemiske målere Salinitetsmåler
This waterproof meter features a unique flat sensor that enables quick, accurate and direct measurements of micro samples as small as 0,12 ml. The sensor does not need to be dipped into the sample, a few drops are sufficient.
  • Auto range change, automatic temperature compensation, automatic standard recognition
  • Automatic hold / auto stable
  • Durable waterproof ABS housing, LCD display with backlight
  • Auto power off after 30 mins, low battery indicator
  • 400 hours battery life (continuous use without backlight)
  • No need for a beaker to calibrate the meter or