Thermo Scientific™ Teleshake 1536-8 Magnetic Microtiter Plate Shaker
Leverandør: THERMO H + P LABORTECHNIK
50101683
HPLT50101683EA 0 NOK
HPLT50101683
Thermo Scientific™ Teleshake 1536-8 Magnetic Microtiter Plate Shaker
Risteapparater Risteapparater, mikroplater
Perform powerful resonance shaking up to 8500 min⁻¹ with Thermo Scientific™ Teleshake 1536-8 Magnetic Microtiter Plate Shakers, which feature a wear-free magnetic drive. They are remote controlled for integrated shaking applications, and are suited for 1536- and 384-well microtiter plates.
  • Acceleration up to 40×g in the resonance range with a minimum of moved masses which translates into little transference of vibrations onto the respective installation areas
  • Easy integration in robotic systems and platform due to small footprint
  • Simultaneous control of up
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Vekt 1,2 kg
B×D×H 103×146×56 mm
Temperaturområde 5...40 °C
Elektronisk 230 V, 50/60 Hz