Safety needles, HENKE-JECT®
Leverandør: HENKE SASS WOLF
8300020446
HSWA8300020446EA 520 NOK
HSWA8300020446
Safety needles, HENKE-JECT®
Sprøytenåler
These safety needles are compatible with HENKE-JECT® syringes as well as all others syringes with Luer and Luer lock according to ISO 594.
  • Safety mechanism is an integrated part of the needle
  • Audible 'click' indicates safety activation
  • Single-handed activation
Sertifiseringer: Conform to EU-Guideline 2010/32/EU and TRBA 250.