Gold chloride løsning 200 mg/dl in deionised water
Leverandør: SIGMA ALDRICH MICROSCOPY
M-Clarity™ quality level = MQ500
HT1004-100ML
HT1004-100MLEA 1070 NOK
HT1004-100ML
Gold chloride løsning 200 mg/dl in deionised water
Gold chloride
Used in procedure HT100 for the toning of sections stained with the silver methanamine procedure.
  • Gold chloride, 0.2% in aqueous solution
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98a1cbeec501f6e31f80564519f57f216ca186e8&vid=8e2d0545dc49fd9bcc3b56122da4478b3923b82e&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance (Colour) Yellow
Appearance (Form) Solution
Appearance (Turbidity)
Essentially free of particles/precipitate
Clear
Performance Conforms