Eosin Y alcoholic solution, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

HT110116-500ML
HT110116-500MLEA 650 NOK
HT110116-500ML
Eosin Y alcoholic solution, Sigma-Aldrich®
Eosin Y

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.

Formel: C₂₀H₆Br₄Na₂O₅
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005040
CAS-nummer: 17372-87-1
ADR: 3,II
IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR